1,3-Propanesultone ≥99%
Supplier: BeanTown Chemical
CAS: 1120-71-4; EC No: 214-317-9; MDL No: MFCD00005355; RTECS: RP5425000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C3H6O3S; MW: 122.14
Melting Point: 30-33°; Boiling Point: 180°/30 mmHg; Flash point: 110°C (230°F)
Density (g/mL): 1.392
Moisture Sensitive
- RTECS: RP5425000
- Moisture Sensitive
- Solid
Formula:
C₃H₆O₃S Boiling Pt: 180 °C/30 mmHg Melting Pt: 30…33 °C Density: 1.392 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00005355 CAS Number: 1120-71-4 EINECS: 214-317-9 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...