Ethyl propiolate ≥99%
Supplier: BeanTown Chemical
CAS: 623-47-2; EC No: 210-795-8; MDL No: MFCD00009184
UN No: UN3272; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: HC≡CCO2CH2CH3; Molecular Formula: C5H6O2; MW: 98.10
Boiling Point: 120°; Flash point: 25°C (77°F)
Density (g/mL): 0.968; Refractive Index: 1.412
- Refractive index: 1.412
- Liquid
Formula:
C₅H₆O₂ Boiling Pt: 120 °C Density: 0.968 g/mL Flash Pt: 25 °C (77 °F) |
MDL Number:
MFCD00009184 CAS Number: 623-47-2 EINECS: 210-795-8 UN: 3272 ADR: 3,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...