Benzophenone ≥99%

Supplier: BeanTown Chemical

Synonyms: Diphenylmethanone, Diphenyl ketone

138280-1KG 138280-250G 138280-5KG
BT138280-1KGEA 68.76 USD
BT138280-1KG BT138280-250G BT138280-5KG
Benzophenone ≥99%
Benzophenone

CAS: 119-61-9; EC No: 204-337-6; MDL No: MFCD00003076; RTECS: DI9950000 UN No: UN3077; Haz Class: 9; Packing Group: III Flakes; Molecular Formula: C13H10O; MW: 182.22 Melting Point: 47-51°; Boiling Point: 305°


  • RTECS: DI9950000
  • Flakes

Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 305 °C
Melting Pt: 47…51 °C
MDL Number: MFCD00003076
CAS Number: 119-61-9
EINECS: 204-337-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR