tert-Butyldimethylsilyl trifluoromethanesulfonate ≥98%

Supplier: BeanTown Chemical

Synonyms: t-Butyldimethylsilyl trifluoromethanesulphonate

138260-25G 138260-5G
BT138260-25GEA 262 USD
BT138260-25G BT138260-5G
tert-Butyldimethylsilyl trifluoromethanesulfonate ≥98%
tert-Butyldimethylsilyl trifluoromethanesulphonate

CAS: 69739-34-0; EC No: 274-102-0; MDL No: MFCD00000405 UN No: UN2920; Haz Class: 8 (3); Packing Group: II Liquid; Linear Formula: CF3SO3Si(CH3)2C(CH3)3; Molecular Formula: C7H15F3O3SSi; MW: 264.34 Boiling Point: 65-67°/12 mmHg; Flash point: 36°C (96°F) Density (g/mL): 1.151; Refractive Index: 1.385 Moisture Sensitive


  • Refractive index: 1.385
  • Moisture Sensitive
  • Liquid

Formula: C₇H₁₅F₃O₃SSi
MW: 264.34 g/mol
Boiling Pt: 65…67 °C/12 mmHg
Density: 1.151 g/mL
Flash Pt: 36 °C (96 °F)
MDL Number: MFCD00000405
CAS Number: 69739-34-0
EINECS: 274-102-0
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR