Calcium oxalate monohydrate ≥99.995%

Supplier: BeanTown Chemical

Synonyms: Oxalic acid calcium salt monohydrate, Calcium ethanedioate monohydrate

138165-25G 138165-5G
BT138165-25GEA 155 USD
BT138165-25G BT138165-5G
Calcium oxalate monohydrate ≥99.995%
Calcium oxalate monohydrate

CAS: 5794-28-5; EC No: 209-260-1; MDL No: MFCD00150039 UN No: UN3288; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: CaC2O4·H2O ; MW: 146.11 Density (g/mL): 2.12 Hygroscopic


  • Hygroscopic
  • Powder

Formula: C₂CaO₄·1H₂O
Density: 2.12 g/mL
MDL Number: MFCD00150039
CAS Number: 5794-28-5
EINECS: 209-260-1
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR