Isohexane (mixture of isomeric branched chain Hexanes) ≥98%

Supplier: BeanTown Chemical

Synonyms: Hexanes

138145-1L 138145-2.5L 138145-500ML
BT138145-1LEA 35 USD
BT138145-1L BT138145-2.5L BT138145-500ML
Isohexane (mixture of isomeric branched chain Hexanes) ≥98%
Isohexane (mixture of isomeric branched chain Hexanes)

CAS: 92112-69-1; EC No: 295-570-2; MDL No: MFCD02179311 UN No: UN1208; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C6H14; MW: 86.18 Melting Point: -95°; Boiling Point: 68-70°; Flash point: -23°C (-9°F) Density (g/mL): 0.672; Refractive Index: 1.379


  • Refractive index: 1.379
  • Liquid

Formula: C₆H₁₄
Boiling Pt: 68…70 °C
Melting Pt: –95 °C
Density: 0.672 g/mL
Flash Pt: –23 °C (–9 °F)
MDL Number: MFCD02179311
CAS Number: 73513-42-5
EINECS: 295-570-2
UN: 1208
ADR: 3,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR