N,N-Dimethylacetamide dimethyl acetal ≥90% stabilised, Technical Grade

Supplier: BeanTown Chemical

Synonyms: (1,1-Dimethoxyethyl)dimethylamine, 1,1-Dimethoxy-N,N-dimethylethanamine

138110-25G 138110-5G
BT138110-25GEA 136 USD
BT138110-25G BT138110-5G
N,N-Dimethylacetamide dimethyl acetal ≥90% stabilised, Technical Grade
N,N-Dimethylacetamide dimethyl acetal

CAS: 18871-66-4; EC No: 242-641-0; MDL No: MFCD00008476 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3C(OCH3)2N(CH3)2; Molecular Formula: C6H15NO2; MW: 133.19 Boiling Point: 118-120°; Flash point: 8°C (46°F) Density (g/mL): 0.911; Refractive Index: 1.411 Moisture Sensitive


  • Refractive index: 1.411
  • Moisture Sensitive
  • Liquid

Formula: C₆H₁₅NO₂
Boiling Pt: 118…120 °C
Density: 0.911 g/mL
Flash Pt: 8 °C (46 °F)
MDL Number: MFCD00008476
CAS Number: 18871-66-4
EINECS: 242-641-0
ADR: 3,II

Order Now

Specification Test Results

Assay 90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR