Aluminium chloride hexahydrate ≥99.9995%
Supplier: BeanTown Chemical
Synonyms:
Aluminum chloride hexahydrate
137960-250G
137960-50G
137960-5G
BT137960-250GEA
308.87
USD
BT137960-250G
BT137960-50G
BT137960-5G
Aluminium chloride hexahydrate ≥99.9995%
Aluminium chloride hexahydrate
CAS: 7784-13-6; EC No: 231-208-1; MDL No: MFCD00149134; RTECS: BD0530000
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline Aggregates; Molecular Formula: AlCl3·6H2O; MW: 241.43
- RTECS: BD0530000
- Crystalline Aggregates
Formula:
AlCl₃·6H₂O MW: 241.43 g/mol Melting Pt: 100 |
MDL Number:
MFCD00149134 CAS Number: 7784-13-6 EINECS: 231-208-1 UN: 3260 ADR: 8 III |
Specification Test Results
Assay | 99.9995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...