Ethyl-4,4-difluoroacetoacetate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Ethyl 4,4-difluoro-3-oxobutanoate, Ethyl 4,4-difluoro-3-oxobutyrate
137945-1G
137945-5G
BT137945-1GEA
88
USD
BT137945-1G
BT137945-5G
Ethyl-4,4-difluoroacetoacetate ≥98%
Ethyl-4,4-difluoroacetoacetate
CAS: 352-24-9; EC No: 206-519-0; MDL No: MFCD00039277
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C6H8F2O3; MW: 166.12
Boiling Point: 162°
Density (g/mL): 1.269; Refractive Index: 1.409
- Refractive index: 1.409
- Liquid
Formula:
C₆H₈F₂O₃ Boiling Pt: 162 °C Density: 1.269 g/mL |
MDL Number:
MFCD00039277 CAS Number: 352-24-9 EINECS: 206-519-0 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...