Cycloheptanone ≥99%

Supplier: BeanTown Chemical

Synonyms: Suberon, Cycloheptan-1-one

137935-250G 137935-50G
BT137935-250GEA 118 USD
BT137935-250G BT137935-50G
Cycloheptanone ≥99%
Cycloheptanone

CAS: 502-42-1; EC No: 207-937-6; MDL No: MFCD00004159; RTECS: GU3325000 UN No: UN1224; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C7H12O; MW: 112.17 Boiling Point: 179°; Flash point: 56°C (133°F) Density (g/mL): 0.951; Refractive Index: 1.461


  • RTECS: GU3325000
  • Refractive index: 1.461
  • Liquid

Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 179 °C
Density: 0.951 g/mL
Flash Pt: 56 °C (133 °F)
MDL Number: MFCD00004159
CAS Number: 502-42-1
EINECS: 207-937-6
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR