Aluminium nitrate nonahydrate 98.0-102.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Aluminum nitrate nonahydrate
137905-1KG
137905-250G
137905-50G
BT137905-1KGEA
211.31
USD
BT137905-1KG
BT137905-250G
BT137905-50G
Aluminium nitrate nonahydrate 98.0-102.0% ACS
Aluminium nitrate nonahydrate
CAS: 7784-27-2; EC No: 236-751-8; MDL No: MFCD00003419; RTECS: BD1050000
UN No: UN1438; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: Al(NO3)3·9H2O ; MW: 375.13
Melting Point: 73°
Hygroscopic
- RTECS: BD1050000
- Hygroscopic
- Crystalline
Formula:
Al(NO₃)₃·9H₂O MW: 375.13 g/mol Melting Pt: 73 °C |
MDL Number:
MFCD00149132 CAS Number: 7784-27-2 EINECS: 236-751-8 UN: 1438 ADR: 5.1,III |
Specification Test Results
Assay | 98.0-102.0% |
Insoluble matter | ≤0.005% |
Chloride (Cl) | ≤0.001% |
Sulfate (SO4) | ≤0.005% |
Calcium (Ca) | ≤0.005% |
Magnesium (Mg) | ≤0.001% |
Potassium (K) | ≤0.002% |
Sodium (Na) | ≤0.005% |
Heavy metals (as Pb) | ≤0.001% |
Iron (Fe) | ≤0.002% |
Substances not precipitated by NH4OH | ≤0.05% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...