Silver(I) sulphide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Disilver sulphide
137855-100G
137855-25G
137855-5G
BT137855-100GEA
421
USD
BT137855-100G
BT137855-25G
BT137855-5G
Silver(I) sulphide ≥99%
Silver(I) sulphide
CAS: 21548-73-2; EC No: 244-438-2; MDL No: MFCD00003406
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Molecular Formula: Ag2S; MW: 247.8
Melting Point: 845°
Density (g/mL): 7.234
- Powder
Formula:
Ag₂S MW: 247.8 g/mol Melting Pt: 842 Density: 7.234 g/mL |
MDL Number:
MFCD00003406 CAS Number: 21548-73-2 EINECS: 244-438-2 ADR: 9 III Merck Index: 12,08677 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...