Antimony(III) chloride ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Antimony trichloride
137845-100G
137845-25G
BT137845-100GEA
153.41
USD
BT137845-100G
BT137845-25G
Antimony(III) chloride ≥99.9%
Antimony(III) chloride
CAS: 10025-91-9; EC No: 233-047-2; MDL No: MFCD00011212; RTECS: CC4900000
UN No: UN1733; Haz Class: 8; Packing Group: II
Crystalline; Linear Formula: SbCl3; MW: 228.12
Melting Point: 73.4°
Moisture Sensitive
- RTECS: CC4900000
- Moisture Sensitive
- Crystalline
Formula:
SbCl₃ MW: 228.12 g/mol Boiling Pt: 223.5 Melting Pt: 73 |
MDL Number:
MFCD00011212 CAS Number: 10025-91-9 EINECS: 233-047-2 UN: 1733 ADR: 8 II Merck Index: 13,00713 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...