1,4-Dibromobutane ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,4-Dibromo butane
137835-100G
137835-500G
BT137835-100GEA
31.29
USD
BT137835-100G
BT137835-500G
1,4-Dibromobutane ≥98%
1,4-Dibromobutane
CAS: 110-52-1; EC No: 203-775-5; MDL No: MFCD00000261; RTECS: EJ7565000
UN No: UN2810; Haz Class: 6.1; Packing Group: III
Liquid; Linear Formula: Br(CH2)4Br; Molecular Formula: C4H8Br2; MW: 215.91
Melting Point: -20°; Boiling Point: 63-65°/6 mmHg; Flash point: 110°C (230°F)
Density (g/mL): 1.808; Refractive Index: 1.519
- RTECS: EJ7565000
- Refractive index: 1.519
- Liquid
Formula:
BrH₂C(CH₂)₂CH₂Br MW: 215.92 g/mol Boiling Pt: 197 Melting Pt: -20 Density: 1.808 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00000261 CAS Number: 110-52-1 EINECS: 203-775-5 UN: 2810 ADR: 6.1 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...