p-Toluenesulphonic acid monohydrate ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate
137825-100G
137825-25G
137825-500G
BT137825-100GEA
18.15
USD
BT137825-100G
BT137825-25G
BT137825-500G
p-Toluenesulphonic acid monohydrate ≥97%
p-Toluenesulphonic acid monohydrate
CAS: 6192-52-5; EC No: 203-180-0; MDL No: MFCD00142137; RTECS: XT6300000
UN No: UN2585; Haz Class: 8; Packing Group: III
Solid; Linear Formula: CH3C6H4SO3H·H2O; Molecular Formula: C7H8O3S·H2O; MW: 190.22
Melting Point: 103-106°; Boiling Point: 140°/22 mmHg; Flash point: 150°C (302°F)
Density (g/mL): 1.24
Hygroscopic
- RTECS: XT6300000
- Hygroscopic
- Solid
Formula:
C₇H₈O₃S·1H₂O Boiling Pt: 140 °C/22 mmHg Melting Pt: 103…106 °C Density: 1.24 g/mL Flash Pt: 150 °C (302 °F) |
MDL Number:
MFCD00142137 CAS Number: 6192-52-5 EINECS: 203-180-0 UN: 2585 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...