Acetonitrile-[D₃] ≥99.8% (Isotopic)
Supplier: BeanTown Chemical
Synonyms:
Cyanomethane-D3, Acetonitrile-d3, Ethannitrile-D3, Trideuteroacetonitrile, Methyl-d3 cyanide, Methyl cyanide-D3, Acetonitrile-[²H₃]
137805-25G
137805-5G
BT137805-25GEA
277.55
USD
BT137805-25G
BT137805-5G
Acetonitrile-[D₃] ≥99.8% (Isotopic)
Acetonitrile-[D₃]
CAS: 2206-26-0; EC No: 218-616-5; MDL No: MFCD00001881
UN No: UN1648; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CD3CN; MW: 44.07
Boiling Point: 80.7°; Flash point: 6°C (43°F)
Density (g/mL): 0.844; Refractive Index: 1.341
- Refractive index: 1.341
- Liquid
Formula:
D₃CCN Boiling Pt: 80.7 °C Density: 0.844 g/mL Flash Pt: 6 °C (43 °F) |
MDL Number:
MFCD00001881 CAS Number: 2206-26-0 EINECS: 218-616-5 UN: 1648 ADR: 3,II |
Specification Test Results
Assay | 99.8% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...