1,5-Diaminopentane ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,5-Pentanediamine, Pentamethylendiamine, Cadaverine, Cadavarine
137770-25G
137770-5G
BT137770-25GEA
384
USD
BT137770-25G
BT137770-5G
1,5-Diaminopentane ≥98%
1,5-Diaminopentane
CAS: 462-94-2; EC No: 207-329-0; MDL No: MFCD00008239
UN No: UN2735; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: C5H14N2; MW: 102.18
Boiling Point: 178-180°; Flash point: 62°C (144°F)
Density (g/mL): 0.873; Refractive Index: 1.458
Air Sensitive , Hygroscopic
- Refractive index: 1.458
- Air Sensitive, Hygroscopic
- Liquid
Formula:
C₅H₁₄N₂ MW: 102.18 g/mol Boiling Pt: 178…180 °C Density: 0.873 g/mL Flash Pt: 62 °C (144 °F) |
MDL Number:
MFCD00008239 CAS Number: 462-94-2 EINECS: 207-329-0 UN: 2735 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...