5-Bromo-2-furaldehyde ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Bromofuran-2-carbaldehyde, 5-Bromofurfural

137740-10G 137740-50G
BT137740-10GEA 102 USD
BT137740-10G BT137740-50G
5-Bromo-2-furaldehyde ≥98%
5-Bromo-2-furaldehyde

CAS: 1899-24-7; MDL No: MFCD00159501 UN No: UN3261; Haz Class: 8; Packing Group: III Needles; Molecular Formula: C5H3BrO2 ; MW: 174.98 Melting Point: 82-85°; Boiling Point: 112°/16 mmHg Air Sensitive


  • Air Sensitive
  • Needles

Formula: C₅H₃BrO₂
MW: 174.98 g/mol
Boiling Pt: 112 °C/16 mmHg
Melting Pt: 82…85 °C
MDL Number: MFCD00159501
CAS Number: 1899-24-7
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR