Antimony(III) fluoride ≥99%

Supplier: BeanTown Chemical

Synonyms: Antimony trifluoride

137735-100G 137735-500G
BT137735-100GEA 58.47 USD
BT137735-100G BT137735-500G
Antimony(III) fluoride ≥99%
Antimony(III) fluoride

CAS: 7783-56-4; EC No: 232-009-2; MDL No: MFCD00011218; RTECS: CC5150000 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III Crystalline; Linear Formula: SbF3; MW: 178.76 Melting Point: 235° Density (g/mL): 4.3 Moisture Sensitive


  • RTECS: CC5150000
  • Moisture Sensitive
  • Crystalline

Formula: F₃Sb
MW: 178.76 g/mol
Melting Pt: 235 °C
Density: 4.3 g/mL
MDL Number: MFCD00011218
CAS Number: 7783-56-4
EINECS: 232-009-2
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR