Ammonium dichromate ≥99.5%

Supplier: BeanTown Chemical

137730-100G 137730-25G
BT137730-100GEA 407 USD
BT137730-100G BT137730-25G
Ammonium dichromate ≥99.5%
Ammonium dichromate

CAS: 7789-09-5; EC No: 232-143-1; MDL No: MFCD00010879; RTECS: HX7650000 UN No: UN1439; Haz Class: 5.1; Packing Group: II Crystalline; Linear Formula: (NH4)2Cr2O7; MW: 252.06 Melting Point: 170° (decomposes)


  • RTECS: HX7650000
  • Crystalline

Formula: (NH₄)₂Cr₂O₇
MW: 252.06 g/mol
Melting Pt: 180 (dec.)
MDL Number: MFCD00010879
CAS Number: 7789-09-5
EINECS: 232-143-1
UN: 1439
ADR: 5.1 II
Merck Index: 13,00516

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR