Manganese(II) chloride, anhydrous ≥97%
Supplier: BeanTown Chemical
CAS: 7773-01-5; EC No: 231-869-6; MDL No: MFCD00011114; RTECS: OO9625000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Crystalline/Flakes; Linear Formula: MnCl2; MW: 125.84
Melting Point: 652°; Boiling Point: 1190°
Density (g/mL): 2.977
Hygroscopic
- RTECS: OO9625000
- Hygroscopic
- Crystalline/Flakes
Formula:
MnCl₂ Boiling Pt: 1190 °C Melting Pt: 652 °C Density: 2.977 g/mL |
MDL Number:
MFCD00011114 CAS Number: 7773-01-5 EINECS: 231-869-6 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...