Aminoacetaldehyde dimethyl acetal ≥99%
Supplier: BeanTown Chemical
Synonyms:
2,2-Dimethoxyethanamine
137720-100G
137720-25G
137720-500G
BT137720-100GEA
122
USD
BT137720-100G
BT137720-25G
BT137720-500G
Aminoacetaldehyde dimethyl acetal ≥99%
Aminoacetaldehyde dimethyl acetal
CAS: 22483-09-6; EC No: 245-026-5; MDL No: MFCD00008135
UN No: UN2733; Haz Class: 3 (8); Packing Group: III
Liquid; Linear Formula: NH2CH2CH(OCH3)2; Molecular Formula: C4H11NO2; MW: 105.14
Boiling Point: 135-139°/95 mmHg; Flash point: 44°C (111°F)
Density (g/mL): 0.965; Refractive Index: 1.417
- Refractive index: 1.417
- Liquid
Formula:
C₄H₁₁NO₂ MW: 105.14 g/mol Boiling Pt: 135…139 °C/95 mmHg Density: 0.965 g/mL Flash Pt: 44 °C (111 °F) |
MDL Number:
MFCD00008135 CAS Number: 22483-09-6 EINECS: 245-026-5 ADR: 3 (8),III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...