2-Chloro-4-fluorophenylacetonitrile ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-(2-Chloro-4-fluorophenyl)acetonitrile

137710-25G 137710-5G
BT137710-25GEA 147 USD
BT137710-25G BT137710-5G
2-Chloro-4-fluorophenylacetonitrile ≥97%
2-Chloro-4-fluorophenylacetonitrile

CAS: 75279-56-0; MDL No: MFCD00236029 UN No: UN3439; Haz Class: 6.1; Packing Group: II Crystalline/Powder; Molecular Formula: C8H5ClFN ; MW: 169.59


  • Crystalline/Powder

Formula: C₈H₅ClFN
MW: 169.59 g/mol
MDL Number: MFCD00236029
CAS Number: 75279-56-0
UN: 3439
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR