Ammonium fluoride hydrate ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
137680-100G
137680-25G
BT137680-100GEA
436.49
USD
BT137680-100G
BT137680-25G
Ammonium fluoride hydrate ≥99.995% (trace metals basis)
Ammonium fluoride hydrate
CAS: 12125-01-8; EC No: 235-185-9; MDL No: MFCD00011423; RTECS: BQ6300000
UN No: UN2505; Haz Class: 6.1; Packing Group: III
Crystalline Aggregates; Linear Formula: NH4F·xH2O ; MW: 37.041 (anhydrous)
Hygroscopic
- RTECS: BQ6300000
- Hygroscopic
- Crystalline Aggregates
Formula:
NH₄F·H₂O MW: 37.04 g/mol (anhy) |
MDL Number:
MFCD00011423 CAS Number: 12125-01-8 EINECS: 235-185-9 UN: 2505 ADR: 6.1 III |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...