Naphthalene ≥99.5%

Supplier: BeanTown Chemical

137615-2KG 137615-500G
BT137615-2KGEA 196 USD
BT137615-2KG BT137615-500G
Naphthalene ≥99.5%
Naphthalene

CAS: 91-20-3; EC No: 202-049-5; MDL No: MFCD00001742; RTECS: QJ0525000 UN No: UN1334; Haz Class: 4.1; Packing Group: III Crystalline; Molecular Formula: C10H8; MW: 128.17 Melting Point: 80-82°; Boiling Point: 218°; Flash point: 78°C (172°F) Density (g/mL): 0.963; Refractive Index: 1.582


  • RTECS: QJ0525000
  • Refractive index: 1.582
  • Crystalline

Formula: C₁₀H₈
MW: 128.17 g/mol
Boiling Pt: 218 °C
Melting Pt: 80…82 °C
Density: 0.963 g/mL
Flash Pt: 78 °C (172 °F)
MDL Number: MFCD00001742
CAS Number: 91-20-3
EINECS: 202-049-5
UN: 1334
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c87c606cb828e0f39bab8b9d50c2c26501e1637d&vid=9e4bc21353014391fc8486208c8757e6375e2ef5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR