Sodium tetrachloroplatinate(II) hydrate ≥99.95% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Disodium tetrachloroplatinate hydrate, Sodium platinum chloride hydrate, Sodium chloroplatinate(II) hydrate

137590-1G 137590-5G
BT137590-1GEA 175.82 USD
BT137590-1G BT137590-5G
Sodium tetrachloroplatinate(II) hydrate ≥99.95% (trace metals basis)
Sodium tetrachloroplatinate(II) hydrate

CAS: 207683-21-4; EC No: 233-051-4; MDL No: MFCD00150188 UN No: UN3288; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: Na2PtCl4·xH2O; MW: 382.88 (anhydrous) Melting Point: 100°


  • Crystalline

Formula: Na₂PtCl₄·xH₂O
MW: 382.88 g/mol (anhy)
Melting Pt: 100 °C
MDL Number: MFCD00150188
CAS Number: 207683-21-4
EINECS: 233-051-4
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR