4-(Dimethylamino)pyridine ≥99%

Supplier: BeanTown Chemical

Synonyms: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine

137520-100G 137520-25G 137520-5G
BT137520-100GEA 173.07 USD
BT137520-100G BT137520-25G BT137520-5G
4-(Dimethylamino)pyridine ≥99%
4-(Dimethylamino)pyridine

CAS: 1122-58-3; EC No: 214-353-5; MDL No: MFCD00006418; RTECS: US9230000 UN No: UN2928; Haz Class: 6.1 (8); Packing Group: I Crystalline; Molecular Formula: C7H10N2; MW: 122.17 Melting Point: 108-110°; Flash point: 110°C (230°F)


  • RTECS: US9230000
  • Crystalline

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 195
Melting Pt: 108…110 °C
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00006418
CAS Number: 1122-58-3
EINECS: 214-353-5
UN: 2928
ADR: 6.1 I

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR