Cyclopropanecarbonitrile ≥98%
Supplier: BeanTown Chemical
Synonyms:
Cyclopropyl cyanide
137460-10G
137460-250G
137460-50G
BT137460-10GEA
38
USD
BT137460-10G
BT137460-250G
BT137460-50G
Cyclopropanecarbonitrile ≥98%
Cyclopropanecarbonitrile
CAS: 5500-21-0; EC No: 226-836-8; MDL No: MFCD00001269
UN No: UN3275; Haz Class: 6.1 (3); Packing Group: II
Liquid; Molecular Formula: C4H5N; MW: 67.09
Boiling Point: 135°; Flash point: 40°C (104°F)
Density (g/mL): 0.911; Refractive Index: 1.421
- Refractive index: 1.421
- Liquid
Formula:
C₄H₅N MW: 67.09 g/mol Boiling Pt: 135 °C Density: 0.911 g/mL Flash Pt: 40 °C (104 °F) |
MDL Number:
MFCD00001269 CAS Number: 5500-21-0 EINECS: 226-836-8 UN: 3275 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...