Zirconium nitride ≥99.5% (trace metals basis excluding Hf)
Supplier: BeanTown Chemical
137405-100G
137405-25G
BT137405-100GEA
274
USD
BT137405-100G
BT137405-25G
Zirconium nitride ≥99.5% (trace metals basis excluding Hf)
Zirconium nitride
CAS: 25658-42-8; EC No: 247-166-2; MDL No: MFCD00049642
UN No: UN3178; Haz Class: 4.1; Packing Group: III
-325 Mesh Powder; Linear Formula: ZrN; MW: 105.23
Melting Point: 2980°
Density (g/mL): 7.09
- -325 Mesh Powder
Formula:
NZr MW: 105.23 g/mol Melting Pt: 2980 °C Density: 7.09 g/mL |
MDL Number:
MFCD00049642 CAS Number: 25658-42-8 EINECS: 247-166-2 UN: 3178 ADR: 4.1,III |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...