Lanthanum(III) trifluoromethanesulphonate, anhydrous ≥99%
Supplier: BeanTown Chemical
Synonyms:
Lanthanum(III) trifluoromethanesulphonate, Lanthanum trifluoromethanesulphonate, Lanthanum triflate
137375-25G
137375-5G
BT137375-25GEA
117
USD
BT137375-25G
BT137375-5G
Lanthanum(III) trifluoromethanesulphonate, anhydrous ≥99%
Lanthanum(III) trifluoromethanesulphonate
CAS: 52093-26-2; MDL No: MFCD00077560
UN No: UN3261; Haz Class: 8; Packing Group: III
Powder; Linear Formula: (CF2CO3)3La; MW: 586.10
Hygroscopic
- Hygroscopic
- Powder
Formula:
C₃F₉LaO₉S₃ MW: 586.12 g/mol |
MDL Number:
MFCD00077560 CAS Number: 52093-26-2 UN: 3261 ADR: 8 III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...