Aluminum phosphate ≥97%

Supplier: BeanTown Chemical

137365-1KG 137365-250G
BT137365-1KGEA 133 USD
BT137365-1KG BT137365-250G
Aluminum phosphate ≥97%
Aluminum phosphate

CAS: 7784-30-7; EC No: 232-056-9; MDL No: MFCD00003430; RTECS: TB6450000 UN No: UN3260; Haz Class: 8; Packing Group: III Powder; Linear Formula: AlPO4; MW: 121.95 Density (g/mL): 2.566


  • RTECS: TB6450000
  • Powder

Formula: AlPO₄
MW: 121.951 g/mol
Density: 2.566 g/mL
MDL Number: MFCD00003430
CAS Number: 7784-30-7
EINECS: 232-056-9
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR