4-Methylpiperidine ≥98%

Supplier: BeanTown Chemical

137350-100ML 137350-500ML
BT137350-100MLEA 31.02 USD
BT137350-100ML BT137350-500ML
4-Methylpiperidine ≥98%
4-Methylpiperidine

CAS: 626-58-4; EC No: 210-954-1; MDL No: MFCD00006005 UN No: UN2733; Haz Class: 3 (8); Packing Group: II Liquid; Molecular Formula: C6H13N; MW: 99.17 Boiling Point: 124°; Flash point: 13°C (55°F) Density (g/mL): 0.838; Refractive Index: 1.446


  • Refractive index: 1.446
  • Liquid

Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 124 °C
Density: 0.838 g/mL
Flash Pt: 13 °C (55 °F)
MDL Number: MFCD00006005
CAS Number: 626-58-4
EINECS: 210-954-1
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR