Selenium tetrabromide ≥99%
Supplier: BeanTown Chemical
137335-25G
137335-5G
BT137335-25GEA
164
USD
BT137335-25G
BT137335-5G
Selenium tetrabromide ≥99%
Selenium tetrabromide
CAS: 7789-65-3; EC No: 232-181-9; MDL No: MFCD00049766
UN No: UN2923; Haz Class: 8 (6.1); Packing Group: II
Powder; Linear Formula: SeBr4; MW: 398.60
Melting Point: 70-80° (decomposes)
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
Br₄Se Melting Pt: 70…80 °C (dec.) |
MDL Number:
MFCD00049766 CAS Number: 7789-65-3 EINECS: 232-181-9 UN: 2923 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...