Iodine monochloride 0.22 N in glacial acetic acid

Supplier: BeanTown Chemical

Synonyms: Iodochloride

137295-1L 137295-4L 137295-500ML
BT137295-1LEA 92.24 USD
BT137295-1L BT137295-4L BT137295-500ML
Iodine monochloride 0.22 N in glacial acetic acid
Iodine monochloride

CAS: 7790-99-0; EC No: 232-236-7; MDL No: MFCD00011354 UN No: UN2920; Haz Class: 8 (3); Packing Group: II Liquid; Linear Formula: ICl; MW: 162.36 Light Sensitive, Moisture Sensitive


  • Light Sensitive, Moisture Sensitive
  • Liquid

Formula: ClI
MW: 162.36 g/mol
MDL Number: MFCD00011354
CAS Number: 7790-99-0
EINECS: 232-236-7
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR