3,5-Bis(trifluoromethyl)styrene ≥98%
Supplier: BeanTown Chemical
137255-1G
137255-5G
BT137255-1GEA
104.44
USD
BT137255-1G
BT137255-5G
3,5-Bis(trifluoromethyl)styrene ≥98%
3,5-Bis(trifluoromethyl)styrene
CAS: 349-59-7; MDL No: MFCD00075538
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C10H6F6; MW: 240.15
Flash point: 50°C (122°F)
Density (g/mL): 1.334
- Liquid
Formula:
C₁₀H₆F₆ MW: 240.15 g/mol Density: 1.334 g/mL Flash Pt: 50 °C (122 °F) |
MDL Number:
MFCD00075538 CAS Number: 349-59-7 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...