Aluminium tri-sec-butanolate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Aluminium Tri-sec-butylate, Aluminum s-butoxide, Aluminium tri-sec-butoxide
137240-100G
137240-2.5KG
137240-500G
BT137240-100GEA
40
USD
BT137240-100G
BT137240-2.5KG
BT137240-500G
Aluminium tri-sec-butanolate ≥97%
Aluminium tri-sec-butanolate
CAS: 2269-22-9; EC No: 218-871-2; MDL No: MFCD00009327
UN No: UN1992; Haz Class: 3 (6.1); Packing Group: III
Liquid; Linear Formula: Al[OCH(CH3)CH2CH3]3; Molecular Formula: C12H27AlO3; MW: 246.32
Boiling Point: 200-206°/30 mmHg; Flash point: 26°C (79°F)
Density (g/mL): 0.967
Moisture Sensitive
- Moisture Sensitive
- Liquid
Formula:
C₁₂H₂₇AlO₃ MW: 246.33 g/mol Boiling Pt: 200…206 °C/30 mmHg Density: 0.967 g/mL Flash Pt: 26 °C (79 °F) |
MDL Number:
MFCD00009327 CAS Number: 2269-22-9 EINECS: 218-871-2 UN: 1992 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...