Zirconium diboride ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

137230-250G 137230-50G
BT137230-250GEA 398.64 USD
BT137230-250G BT137230-50G
Zirconium diboride ≥99.5% (trace metals basis)
Zirconium diboride

CAS: 12045-64-6; EC No: 234-963-5; MDL No: MFCD00064648 UN No: UN3178; Haz Class: 4.1; Packing Group: III -325 Mesh Powder; Linear Formula: ZrB2; MW: 112.84 Melting Point: 3000°


  • -325 Mesh Powder

Formula: B₂Zr
MW: 112.84 g/mol
Melting Pt: 3000 °C
MDL Number: MFCD00064648
CAS Number: 12045-64-6
EINECS: 234-963-5
UN: 3178
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR