Cobalt naphthenate 50% (w/w) in mineral spirits

Supplier: BeanTown Chemical

Synonyms: Naphthenic Acid Cobalt Salt

137165-100G 137165-2KG 137165-500G
BT137165-100GEA 59.81 USD
BT137165-100G BT137165-2KG BT137165-500G
Cobalt naphthenate 50% (w/w) in mineral spirits
Cobalt naphthenate

CAS: 61789-51-3; EC No: 263-064-0; MDL No: MFCD02094763; RTECS: QK8925000 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid Flash point: 40°C (104°F)


  • RTECS: QK8925000
  • Liquid

Formula: C₂₂H₁₄CoO₄
MW: 401.28 g/mol
Flash Pt: 40 °C (104 °F)
MDL Number: MFCD02094763
CAS Number: 61789-51-3
EINECS: 263-064-0
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR