4-Fluorobenzoyl chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: p-Fluorobenzoyl chloride

137095-100G 137095-25G 137095-500G
BT137095-100GEA 135 USD
BT137095-100G BT137095-25G BT137095-500G
4-Fluorobenzoyl chloride ≥98%
4-Fluorobenzoyl chloride

CAS: 403-43-0; EC No: 206-961-4; MDL No: MFCD00000684 UN No: UN3094; Haz Class: 8 (4.3); Packing Group: II Liquid; Molecular Formula: C7H4ClFO ; MW: 158.56 Melting Point: 10-12°; Boiling Point: 82°/20 mmHg; Flash point: 82°C (180°F) Density (g/mL): 1.342; Refractive Index: 1.532 Moisture Sensitive


  • Refractive index: 1.532
  • Moisture Sensitive
  • Liquid

Formula: C₇H₄ClFO
Boiling Pt: 82 °C/20 mmHg
Melting Pt: 10…12 °C
Density: 1.342 g/mL
Flash Pt: 82 °C (180 °F)
MDL Number: MFCD00000684
CAS Number: 403-43-0
EINECS: 206-961-4
UN: 3094
ADR: 8,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR