Titanium(IV) methoxide ≥95% may contain up to 5% methanol

Supplier: BeanTown Chemical

Synonyms: Titanium tetramethanolate, Titanium (IV) methoxide

137085-100G 137085-25G 137085-5G
BT137085-100GEA 189 USD
BT137085-100G BT137085-25G BT137085-5G
Titanium(IV) methoxide ≥95% may contain up to 5% methanol
Titanium(IV) methoxide

CAS: 992-92-7; EC No: 213-596-4; MDL No: MFCD00014883 UN No: UN3181; Haz Class: 4.1; Packing Group: III Granular/Lump; Linear Formula: Ti(OCH3)4; MW: 172.00 Melting Point: 200-210°; Boiling Point: 243°/52 mmHg; Flash point: 14°C (57°F) Moisture Sensitive


  • Moisture Sensitive
  • Granular/Lump

Formula: C₄H₁₂O₄Ti
MW: 172.02 g/mol
Boiling Pt: 243 °C/52 mmHg
Melting Pt: 200…210 °C
Flash Pt: 14 °C (57 °F)
MDL Number: MFCD00014883
CAS Number: 992-92-7
EINECS: 213-596-4
UN: 3181
ADR: 4.1 III

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR