Cyclobutanone ≥98% stabilized

Supplier: BeanTown Chemical

137035-1G 137035-25G 137035-5G
BT137035-1GEA 79 USD
BT137035-1G BT137035-25G BT137035-5G
Cyclobutanone ≥98% stabilized
Cyclobutanone

CAS: 1191-95-3; EC No: 214-745-6; MDL No: MFCD00001332 UN No: UN1224 ; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C4H6O; MW: 70.09 Boiling Point: 99°; Flash point: 27°C (81°F) Density (g/mL): 0.938; Refractive Index: 1.421


  • Refractive index: 1.421
  • Liquid

Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 99 °C
Density: 0.938 g/mL
Flash Pt: 27 °C (81 °F)
MDL Number: MFCD00001332
CAS Number: 1191-95-3
EINECS: 214-745-6
UN: 1224
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR