2-Hydroxypyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone

137020-100G 137020-25G 137020-500G
BT137020-100GEA 112 USD
BT137020-100G BT137020-25G BT137020-500G
2-Hydroxypyridine ≥98%
2-Hydroxypyridine

CAS: 142-08-5; EC No: 205-520-3; MDL No: MFCD00006268; RTECS: UV1144050 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Molecular Formula: C5H5NO; MW: 95.10 Melting Point: 105-107°; Boiling Point: 280-281°


  • RTECS: UV1144050
  • Crystalline/Powder

Formula: C₅H₅NO
MW: 95.10 g/mol
Boiling Pt: 280…281 °C
Melting Pt: 105…107 °C
MDL Number: MFCD00006268
CAS Number: 142-08-5
EINECS: 205-520-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR