Paraformaldehyde ≥97%

Supplier: BeanTown Chemical

136950-100G 136950-2.5KG 136950-500G
BT136950-100GEA 32.56 USD
BT136950-100G BT136950-2.5KG BT136950-500G
Paraformaldehyde ≥97%
Paraformaldehyde

CAS: 30525-89-4; EC No: 200-001-8; MDL No: MFCD00133991; RTECS: RV0540000 UN No: UN2213; Haz Class: 4.1; Packing Group: III Powder; Linear Formula: (CH2O)n; MW: 30.03 (monomer) Melting Point: 120-170° (decomposes); Flash point: 70°C (158°F) Density (g/mL): 0.88 Moisture Sensitive


  • RTECS: RV0540000
  • Moisture Sensitive
  • Powder

Formula: (CH₂O)n
Melting Pt: 120…170 °C (dec.)
Density: 0.88 g/mL
Flash Pt: 70 °C (158 °F)
MDL Number: MFCD00133991
CAS Number: 30525-89-4
EINECS: 200-001-8
UN: 2213
ADR: 4.1,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR