Barium titanate ≥99% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide
136930-2KG
136930-500G
136930-50G
BT136930-2KGEA
136
USD
BT136930-2KG
BT136930-500G
BT136930-50G
Barium titanate ≥99% (trace metals basis)
Barium titanate
CAS: 12047-27-7; EC No: 234-975-0; MDL No: MFCD00003447
UN No: UN1564; Haz Class: 6.1; Packing Group: III
-325 Mesh Powder ; Molecular Formula: BaTiO3; MW: 233.19
Density (g/mL): 6.08
- -325 Mesh Powder
Formula:
BaO₃Ti MW: 233.21 g/mol Density: 6.08 g/mL |
MDL Number:
MFCD00003447 CAS Number: 12047-27-7 EINECS: 234-975-0 UN: 1564 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...