tert-Butylchlorodiphenylsilane ≥97%
Supplier: BeanTown Chemical
Synonyms:
tert-Butyldiphenylchlorosilane, tert-Butyl-diphenylchlorosilane
136780-10G
136780-250G
136780-50G
BT136780-10GEA
46
USD
BT136780-10G
BT136780-250G
BT136780-50G
tert-Butylchlorodiphenylsilane ≥97%
tert-Butylchlorodiphenylsilane
CAS: 58479-61-1; EC No: 261-282-0; MDL No: MFCD00000497
UN No: UN2987; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: (CH3)3CSi(C6H5)2Cl; Molecular Formula: C16H19ClSi; MW: 274.87
Boiling Point: 90°/0.01 mmHg; Flash point: 112°C (234°F)
Density (g/mL): 1.057; Refractive Index: 1.568
Moisture Sensitive
- Refractive index: 1.568
- Moisture Sensitive
- Liquid
Formula:
C₁₆H₁₉ClSi MW: 274.87 g/mol Boiling Pt: 90 °C/0.01 mmHg Density: 1.057 g/mL Flash Pt: 112 °C (234 °F) |
MDL Number:
MFCD00000497 CAS Number: 58479-61-1 EINECS: 261-282-0 UN: 2987 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...