Barium bromide dihydrate ≥99%

Supplier: BeanTown Chemical

136765-100G 136765-500G
BT136765-100GEA 54 USD
BT136765-100G BT136765-500G
Barium bromide dihydrate ≥99%
Barium bromide dihydrate

CAS: 7791-28-8; EC No: 234-140-0; MDL No: MFCD00149153 UN No: UN1564; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: BaBr2·2H2O; MW: 333.17


  • Crystalline

Formula: BaBr₂·2H₂O
MW: 333.17 g/mol
MDL Number: MFCD00149153
CAS Number: 7791-28-8
EINECS: 234-140-0
UN: 1564
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR