3-Fluoro-4-nitrobenzyl bromide ≥97%
Supplier: BeanTown Chemical
136715-1G
136715-5G
BT136715-1GEA
203.27
USD
BT136715-1G
BT136715-5G
3-Fluoro-4-nitrobenzyl bromide ≥97%
3-Fluoro-4-nitrobenzyl bromide
CAS: 131858-37-2; MDL No: MFCD03094237
UN No: UN3261; Haz Class: 8; Packing Group: II
Solid; Molecular Formula: C7H5BrFNO2; MW: 234.02
Melting Point: 46-48°
- Solid
Formula:
C₇H₅BrFNO₂ Melting Pt: 46…48 °C |
MDL Number:
MFCD03094237 CAS Number: 131858-37-2 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...