Maleimide ≥98%
Supplier: BeanTown Chemical
CAS: 541-59-3; EC No: 208-787-4; MDL No: MFCD00005494; RTECS: ON4800000
UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III
Crystalline; Molecular Formula: C4H3NO2; MW: 97.07
Melting Point: 91-93°
Density (g/mL): 1.249
- RTECS: ON4800000
- Crystalline
Formula:
C₄H₃NO₂ Melting Pt: 91…93 °C Density: 1.249 g/mL |
MDL Number:
MFCD00005494 CAS Number: 541-59-3 EINECS: 208-787-4 UN: 2923 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...