Silver(II) oxide ≥99.9%

Supplier: BeanTown Chemical

136670-100G 136670-25G 136670-5G
BT136670-100GEA 768 USD
BT136670-100G BT136670-25G BT136670-5G
Silver(II) oxide ≥99.9%
Silver(II) oxide

CAS: 1301-96-8; EC No: 215-098-2; MDL No: MFCD00044285 UN No: UN3085; Haz Class: 5.1 (8); Packing Group: II -100 Mesh Powder; Molecular Formula: AgO; MW: 123.87 Melting Point: 100° (decomposes) Density (g/mL): 7.483 Light Sensitive, Moisture Sensitive


  • Light Sensitive, Moisture Sensitive
  • -100 Mesh Powder

Formula: AgO
MW: 123.87
Melting Pt: 100 (dec.)
Density: 7.483 g/mL
MDL Number: MFCD00044285
CAS Number: 1301-96-8
EINECS: 215-098-2
UN: 3085
ADR: 5.1 II
Merck Index: 12,08666

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR