Sodium sulfide hydrate ≥90%, Technical Grade

Supplier: BeanTown Chemical
Danger

Synonyms: di-Sodium sulphide hydrate

136640-2KG 136640-500G
BT136640-2KGEA 97.74 USD
BT136640-2KG BT136640-500G
Sodium sulfide hydrate ≥90%, Technical Grade
Sodium sulphide hydrate

CAS: 27610-45-3; EC No: 215-211-5; MDL No: MFCD00149183 UN No: UN1849; Haz Class: 8; Packing Group: II Flakes; Molecular Formula: Na2S·xH2O; MW: 78.04 (anhydrous) Melting Point: 50° (decomposes) Density (g/mL): 1.427 Hygroscopic

Formula: Na₂S·xH₂O
MW: 96.06 g/mol (anhy)
Melting Pt: 50 °C (dec.)
Density: 1.427
Storage Temperature: Refrigerator
MDL Number: MFCD00149183
CAS Number: 27610-45-3
EINECS: 215-211-5
UN: 1849
ADR: 8,II

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR